Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183300.2 | NP_899123.2 | 5 - 6 | 639 | 73 | |
GenBank mRNA | DQ303413.1 | - | 5 - 6 | 639 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |