Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kank4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172872.3 | NP_766460.2 | 3 - 4 | 2127 | 128 | |
GenBank mRNA | AK081525.1 | - | 3 - 4 | 2065 | 128 | |
BC060737.1 | - | 3 - 4 | 2141 | 4191502 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |