Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otop3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027132.2 | NP_081408.2 | 2 - 3 | 583 | 54 | |
GenBank mRNA | AB087505.1 | - | 2 - 3 | 538 | 54 | |
AK043266.1 | - | 2 - 3 | 538 | 54 | ||
BC145613.1 | - | 2 - 3 | 500 | 9054179 | 54 | |
BC150803.1 | - | 2 - 3 | 500 | 9087714 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |