Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252614.1 | NP_001239543.1 | 4 - 5 | 369 | 89 | |
NM_016893.5 | NP_058589.2 | 3 - 4 | 280 | 89 | ||
NR_045554.1 | 3 - 4 | 699 | 89 | |||
GenBank mRNA | AK048520.1 | - | 3 - 4 | 418 | 89 | |
AK051811.1 | - | 3 - 4 | 237 | 89 | ||
AK134537.1 | - | 3 - 4 | 373 | 89 | ||
AK163494.1 | - | 3 - 4 | 396 | 89 | ||
BC010666.1 | - | 4 - 5 | 348 | 3594582 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |