Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011155.2 | NP_035285.2 | 1 - 2 | 222 | 73 | |
GenBank mRNA | AF018262.1 | - | 1 - 2 | 193 | 73 | |
BB868382.1 | - | 1 - 2 | 203 | 73 | ||
BC003744.1 | - | 1 - 2 | 178 | 3590322 | 73 | |
DV653678.1 | - | 1 - 2 | 159 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |