Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pyroxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183165.3 | NP_898988.2 | 10 - 11 | 1151 | 52 | |
GenBank mRNA | AK165677.1 | - | 10 - 11 | 1154 | 52 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |