Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178884.5 | NP_849215.3 | 13 - 14 | 4562 | 56 | |
GenBank mRNA | AK029863.1 | - | 13 - 14 | 4351 | 56 | |
AK122340.1 | - | 6 - 7 | 2800 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |