Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178884.5 | NP_849215.3 | 15 - 16 | 4945 | 62 | |
GenBank mRNA | AK029863.1 | - | 15 - 16 | 4734 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |