Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008720.2 | NP_032746.2 | 3 - 4 | 434 | 118 | |
GenBank mRNA | AK154282.1 | - | 3 - 4 | 354 | 118 | |
AK155379.1 | - | 3 - 4 | 354 | 118 | ||
BC052437.1 | - | 3 - 4 | 434 | 6405214 | 118 | |
BC054539.1 | - | 3 - 4 | 434 | 6405214 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |