Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081076.2 | NP_001074545.1 | 12 - 13 | 1996 | 60 | |
GenBank mRNA | AY395631.1 | - | 12 - 13 | 1996 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |