Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005784.1 | NP_001005784.1 | 19 - 20 | 2579 | 94 | |
NM_172696.2 | NP_766284.2 | 19 - 20 | 2579 | 94 | ||
GenBank mRNA | AK044043.1 | - | 1 - 2 | 87 | 94 | |
AK077268.1 | - | 6 - 7 | 861 | 94 | ||
AK147866.1 | - | 19 - 20 | 2528 | 94 | ||
AK153597.1 | - | 8 - 9 | 1434 | 94 | ||
AK156166.1 | - | 8 - 9 | 1047 | 94 | ||
BC062194.1 | - | 7 - 8 | 881 | 94 | ||
BC082787.1 | - | 8 - 9 | 972 | 94 | ||
BC141404.1 | - | 19 - 20 | 2555 | 9056017 | 94 | |
CD547724.1 | - | 3 - 4 | 451 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |