Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172696.2 | NP_766284.2 | 22 - 23 | 3283 | 62 | |
GenBank mRNA | AK050545.1 | - | 18 - 19 | 2259 | 62 | |
AK077268.1 | - | 9 - 10 | 1565 | 62 | ||
AK147866.1 | - | 22 - 23 | 3232 | 62 | ||
BC082787.1 | - | 11 - 12 | 1676 | 62 | ||
BC141404.1 | - | 22 - 23 | 3259 | 9056017 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |