Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005784.1 | NP_001005784.1 | 12 - 13 | 1675 | 104 | |
NM_172696.2 | NP_766284.2 | 12 - 13 | 1675 | 104 | ||
GenBank mRNA | AK050545.1 | - | 12 - 13 | 1647 | 104 | |
AK147866.1 | - | 12 - 13 | 1624 | 104 | ||
AK153597.1 | - | 1 - 2 | 530 | 104 | ||
AK156166.1 | - | 1 - 2 | 143 | 104 | ||
BC050846.1 | - | 12 - 13 | 1609 | 104 | ||
BC141404.1 | - | 12 - 13 | 1651 | 9056017 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |