Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900015382.mm.1 |
NM_001033172.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015382.mm.1 |
NM_001033172.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015382.mm.1 |
TC1900001739.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900015384.mm.1 |
NM_001033172.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015384.mm.1 |
NM_001033172.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015384.mm.1 |
TC1900001739.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900015383.mm.1 |
NM_001033172.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015383.mm.1 |
NM_001033172.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015383.mm.1 |
TC1900001739.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900015381.mm.1 |
NM_001033172.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015381.mm.1 |
NM_001033172.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015381.mm.1 |
NM_001164367.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015381.mm.1 |
NM_001164367.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015381.mm.1 |
TC1900001739.mm.1 |
MTA TC |