Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164583.1 | NP_001158055.1 | 17 - 18 | 2704 | 91 | |
NM_001164584.1 | NP_001158056.1 | 17 - 18 | 2649 | 91 | ||
NM_001164585.1 | NP_001158057.1 | 18 - 19 | 2714 | 91 | ||
NM_198412.2 | NP_940804.1 | 18 - 19 | 2623 | 91 | ||
GenBank mRNA | AK049935.1 | - | 5 - 6 | 666 | 91 | |
AK122293.1 | - | 17 - 18 | 2625 | 91 | ||
AK147570.1 | - | 18 - 19 | 2586 | 91 | ||
AK147637.1 | - | 18 - 19 | 2586 | 91 | ||
AK163657.1 | - | 17 - 18 | 2523 | 91 | ||
BC035262.1 | - | 12 - 13 | 1858 | 91 | ||
BC060734.1 | - | 17 - 18 | 2704 | 6409942 | 91 | |
BC064460.1 | - | 18 - 19 | 2592 | 6821822 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |