Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030094.1 | NP_084370.1 | 3 - 4 | 436 | 59 | |
GenBank mRNA | AK019109.1 | - | 3 - 4 | 441 | 59 | |
AK134929.1 | - | 3 - 4 | 424 | 59 | ||
AK141256.1 | - | 3 - 4 | 424 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |