Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lix1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163170.1 | NP_001156642.1 | 3 - 4 | 645 | 103 | |
GenBank mRNA | AK051276.1 | - | 3 - 4 | 630 | 103 | |
AK143438.1 | - | 3 - 4 | 654 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |