Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr3gl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293701.1 | NP_001280630.1 | 4 - 5 | 353 | 83 | |
NM_027241.4 | NP_081517.2 | 5 - 6 | 521 | 83 | ||
GenBank mRNA | AK008069.1 | - | 5 - 6 | 487 | 83 | |
BC027083.1 | - | 5 - 6 | 471 | 5357914 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |