Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpy19l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178704.3 | NP_848819.2 | 2 - 3 | 343 | 131 | |
GenBank mRNA | AF538322.1 | - | 2 - 3 | 327 | 131 | |
AK031345.1 | - | 2 - 3 | 335 | 131 | ||
AK033395.1 | - | 2 - 3 | 300 | 131 | ||
AK034217.1 | - | 2 - 3 | 344 | 131 | ||
AK045758.1 | - | 2 - 3 | 244 | 131 | ||
BC050065.1 | - | 2 - 3 | 287 | 30016211 | 131 | |
BC059897.1 | - | 2 - 3 | 240 | 6821433 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |