Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026360.3 | NP_080636.2 | 3 - 4 | 402 | 116 | |
GenBank mRNA | AK010310.1 | - | 3 - 4 | 395 | 116 | |
AK076982.1 | - | 3 - 4 | 387 | 116 | ||
BC060168.1 | - | 3 - 4 | 382 | 116 | ||
BC095944.1 | - | 3 - 4 | 379 | 4935970 | 116 | |
BC132242.1 | - | 3 - 4 | 394 | 40130519 | 116 | |
BC132244.1 | - | 3 - 4 | 394 | 40130521 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |