Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026360.3 | NP_080636.2 | 7 - 8 | 779 | 108 | |
GenBank mRNA | AK010310.1 | - | 7 - 8 | 772 | 108 | |
AK076982.1 | - | 7 - 8 | 764 | 108 | ||
AK169595.1 | - | 5 - 6 | 402 | 108 | ||
BC060168.1 | - | 7 - 8 | 759 | 108 | ||
BC095944.1 | - | 7 - 8 | 756 | 4935970 | 108 | |
BC132242.1 | - | 7 - 8 | 771 | 40130519 | 108 | |
BC132244.1 | - | 7 - 8 | 771 | 40130521 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |