Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253847.1 | NP_001240776.1 | 2 - 3 | 259 | 89 | |
GenBank mRNA | AF167441.1 | - | 2 - 3 | 259 | 89 | |
AK051737.1 | - | 2 - 3 | 198 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |