Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163126.1 | NP_001156598.1 | 19 - 20 | 2193 | 69 | |
GenBank mRNA | AK030656.1 | - | 19 - 20 | 2194 | 69 | |
CX240209.1 | - | 2 - 3 | 333 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |