Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168491.1 | NP_001161963.1 | 4 - 5 | 643 | 100 | |
NM_001168492.1 | NP_001161964.1 | 4 - 5 | 643 | 100 | ||
NM_011050.4 | NP_035180.2 | 4 - 5 | 731 | 100 | ||
GenBank mRNA | AK134366.1 | - | 4 - 5 | 732 | 100 | |
AK172654.1 | - | 4 - 5 | 599 | 100 | ||
BC055739.1 | - | 4 - 5 | 578 | 6414089 | 100 | |
CT010188.1 | - | 3 - 4 | 441 | 100 | ||
D50465.1 | - | 3 - 4 | 511 | 100 | ||
D86344.1 | - | 4 - 5 | 527 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |