Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010123.3 | NP_034253.3 | 1 - 2 | 223 | 117 | |
GenBank mRNA | AK082006.1 | - | 1 - 2 | 189 | 117 | |
AK145132.1 | - | 1 - 2 | 231 | 117 | ||
AK145629.1 | - | 1 - 2 | 188 | 117 | ||
AK149482.1 | - | 1 - 2 | 176 | 117 | ||
AK161634.1 | - | 1 - 2 | 180 | 117 | ||
AK167010.1 | - | 1 - 2 | 190 | 117 | ||
AK172899.1 | - | 1 - 2 | 160 | 117 | ||
U14172.1 | - | 1 - 2 | 173 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus 1700 Probe spans exons |