Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029264.2 | NP_083540.1 | 6 - 7 | 659 | 70 | |
GenBank mRNA | AK014686.1 | - | 4 - 5 | 956 | 70 | |
AK016396.1 | - | 3 - 4 | 248 | 70 | ||
AK133079.1 | - | 6 - 7 | 660 | 70 | ||
AM690754.1 | - | 3 - 4 | 543 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |