Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pggt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172627.3 | NP_766215.1 | 3 - 4 | 351 | 89 | |
GenBank mRNA | AK028188.1 | - | 3 - 4 | 209 | 89 | |
AK081731.1 | - | 3 - 4 | 352 | 89 | ||
BC141021.1 | - | 3 - 4 | 349 | 9055633 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |