Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tas1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031867.2 | NP_114073.1 | 4 - 5 | 1577 | 107 | |
GenBank mRNA | AF301161.1 | - | 4 - 5 | 1473 | 107 | |
AF301162.1 | - | 4 - 5 | 1473 | 107 | ||
AF337040.1 | - | 4 - 5 | 1655 | 107 | ||
AK153579.1 | - | 4 - 5 | 1527 | 107 | ||
AY032622.1 | - | 4 - 5 | 1473 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Ion Channel Detect Genomic DNA Mus musculus Probe spans exons |