Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175152.4 | NP_780361.2 | 3 - 4 | 396 | 82 | |
GenBank mRNA | AK008225.1 | - | 3 - 4 | 368 | 82 | |
AK038184.1 | - | 3 - 4 | 370 | 82 | ||
AK050740.1 | - | 3 - 4 | 410 | 82 | ||
BC094395.1 | - | 3 - 4 | 377 | 5700290 | 82 | |
BC115485.1 | - | 3 - 4 | 394 | 82 | ||
BC115486.1 | - | 3 - 4 | 394 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |