Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK020079.1 | - | 2 - 3 | 276 | 70 | |
AK042581.1 | - | 2 - 3 | 680 | 70 | ||
AK046761.1 | - | 2 - 3 | 685 | 70 | ||
AK047599.1 | - | 2 - 3 | 709 | 70 | ||
AK050948.1 | - | 2 - 3 | 692 | 70 | ||
AK079935.1 | - | 2 - 3 | 693 | 70 | ||
AK157398.1 | - | 2 - 3 | 692 | 70 | ||
AK157956.1 | - | 3 - 4 | 597 | 70 | ||
AK164493.1 | - | 2 - 3 | 703 | 70 | ||
AK172043.1 | - | 2 - 3 | 698 | 70 | ||
BC024877.1 | - | 2 - 3 | 696 | 70 | ||
BC051109.1 | - | 2 - 3 | 665 | 70 | ||
BC096599.1 | - | 2 - 3 | 684 | 70 | ||
BC125432.1 | - | 2 - 3 | 691 | 40129947 | 70 | |
BC132099.1 | - | 2 - 3 | 691 | 40130376 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |