Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK014568.1 | - | 7 - 8 | 1302 | 96 | |
AK042581.1 | - | 7 - 8 | 1288 | 96 | ||
AK046761.1 | - | 7 - 8 | 1293 | 96 | ||
AK047599.1 | - | 5 - 6 | 1945 | 96 | ||
AK050948.1 | - | 7 - 8 | 1300 | 96 | ||
AK079935.1 | - | 7 - 8 | 1301 | 96 | ||
AK157398.1 | - | 7 - 8 | 1300 | 96 | ||
AK157956.1 | - | 8 - 9 | 1205 | 96 | ||
AK164493.1 | - | 7 - 8 | 1311 | 96 | ||
AK172043.1 | - | 7 - 8 | 1306 | 96 | ||
AK179132.1 | - | 1 - 2 | 58 | 96 | ||
BC024877.1 | - | 7 - 8 | 1304 | 96 | ||
BC051109.1 | - | 6 - 7 | 1185 | 96 | ||
BC096599.1 | - | 6 - 7 | 1204 | 96 | ||
BC125432.1 | - | 7 - 8 | 1299 | 40129947 | 96 | |
BC132099.1 | - | 7 - 8 | 1299 | 40130376 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |