Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177995.1 | NP_001171466.1 | 11 - 12 | 2976 | 144 | |
NM_001291026.1 | NP_001277955.1 | 11 - 12 | 2970 | 144 | ||
NM_001291029.1 | NP_001277958.1 | 11 - 12 | 2979 | 144 | ||
NM_027504.3 | NP_081780.3 | 11 - 12 | 2973 | 144 | ||
GenBank mRNA | AB078338.1 | - | 11 - 12 | 2865 | 144 | |
AK083829.1 | - | 11 - 12 | 2861 | 144 | ||
BC053082.1 | - | 11 - 12 | 2910 | 144 | ||
BC059838.1 | - | 11 - 12 | 2957 | 6409778 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |