Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177995.1 | NP_001171466.1 | 8 - 9 | 1306 | 66 | |
NM_001291026.1 | NP_001277955.1 | 8 - 9 | 1300 | 66 | ||
NM_001291029.1 | NP_001277958.1 | 8 - 9 | 1306 | 66 | ||
NM_027504.3 | NP_081780.3 | 8 - 9 | 1303 | 66 | ||
GenBank mRNA | AB078338.1 | - | 8 - 9 | 1195 | 66 | |
AK083829.1 | - | 8 - 9 | 1190 | 66 | ||
BC053082.1 | - | 8 - 9 | 1241 | 66 | ||
BC059838.1 | - | 8 - 9 | 1284 | 6409778 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |