Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrkh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028307.1 | NP_082583.1 | 11 - 12 | 1615 | 83 | |
GenBank mRNA | BC049363.1 | - | 11 - 12 | 1615 | 6490411 | 83 |
BC057030.1 | - | 11 - 12 | 1631 | 6833743 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |