Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146196.1 | NP_001139668.1 | 14 - 15 | 2071 | 81 | |
NM_009132.2 | NP_033158.2 | 12 - 13 | 1771 | 81 | ||
GenBank mRNA | AK133484.1 | - | 14 - 15 | 2027 | 81 | |
AK136080.1 | - | 12 - 13 | 1613 | 81 | ||
AK167739.1 | - | 14 - 15 | 2026 | 81 | ||
AK171530.1 | - | 14 - 15 | 2057 | 81 | ||
AK173295.1 | - | 14 - 15 | 2033 | 81 | ||
BC063328.1 | - | 14 - 15 | 2026 | 30314464 | 81 | |
U04354.1 | - | 14 - 15 | 2026 | 81 | ||
Y13971.1 | - | 12 - 13 | 1726 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |