Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025776.2 | NP_080052.1 | 1 - 2 | 95 | 68 | |
GenBank mRNA | AK018452.1 | - | 1 - 2 | 118 | 68 | |
AK045116.1 | - | 1 - 2 | 115 | 68 | ||
AK150271.1 | - | 1 - 2 | 108 | 68 | ||
AK151758.1 | - | 1 - 2 | 108 | 68 | ||
AK152252.1 | - | 1 - 2 | 108 | 68 | ||
AK165277.1 | - | 1 - 2 | 113 | 68 | ||
AK166600.1 | - | 1 - 2 | 116 | 68 | ||
AK167509.1 | - | 1 - 2 | 135 | 68 | ||
AK168919.1 | - | 1 - 2 | 96 | 68 | ||
BC042691.1 | - | 1 - 2 | 114 | 68 | ||
BC080205.1 | - | 1 - 2 | 93 | 3709760 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |