Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025776.2 | NP_080052.1 | 3 - 4 | 179 | 100 | |
GenBank mRNA | AK018452.1 | - | 3 - 4 | 202 | 100 | |
AK045116.1 | - | 3 - 4 | 199 | 100 | ||
AK150271.1 | - | 3 - 4 | 192 | 100 | ||
AK151758.1 | - | 3 - 4 | 192 | 100 | ||
AK152252.1 | - | 3 - 4 | 192 | 100 | ||
AK165277.1 | - | 3 - 4 | 197 | 100 | ||
AK166600.1 | - | 3 - 4 | 200 | 100 | ||
AK167509.1 | - | 3 - 4 | 219 | 100 | ||
AK168919.1 | - | 3 - 4 | 180 | 100 | ||
BC042691.1 | - | 3 - 4 | 198 | 100 | ||
BC080205.1 | - | 3 - 4 | 177 | 3709760 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |