Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081073.1 | NP_001074542.1 | 10 - 11 | 1789 | 99 | |
GenBank mRNA | AK035879.1 | - | 9 - 10 | 1569 | 99 | |
BC119216.1 | - | 10 - 11 | 1670 | 8733965 | 99 | |
BC119218.1 | - | 10 - 11 | 1670 | 8733967 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |