Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271678.1 | NP_001258607.1 | 5 - 6 | 1241 | 123 | |
NM_001271679.1 | NP_001258608.1 | 5 - 6 | 1241 | 123 | ||
GenBank mRNA | JQ247189.1 | - | 5 - 6 | 1241 | 123 | |
JQ247190.1 | - | 5 - 6 | 1241 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Mus musculus Probe spans exons |