Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023718.3 | NP_076207.1 | 6 - 7 | 761 | 100 | |
GenBank mRNA | AF203701.1 | - | 6 - 7 | 765 | 100 | |
AF213260.1 | - | 6 - 7 | 765 | 100 | ||
AK142587.1 | - | 5 - 6 | 619 | 100 | ||
AK143970.1 | - | 6 - 7 | 762 | 100 | ||
BC071214.1 | - | 5 - 6 | 611 | 5042082 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |