Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177772.1 | NP_001171243.1 | 7 - 8 | 1161 | 86 | |
NM_021471.2 | NP_067446.1 | 7 - 8 | 1161 | 86 | ||
GenBank mRNA | AK045782.1 | - | 1164 | 86 | ||
AK048926.1 | - | 7 - 8 | 1131 | 86 | ||
AK161010.1 | - | 1131 | 86 | |||
AY007379.1 | - | 7 - 8 | 997 | 86 | ||
BC078456.1 | - | 7 - 8 | 1111 | 30360663 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |