Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020495.1 | NP_065241.1 | 7 - 8 | 805 | 105 | |
GenBank mRNA | AB031959.1 | - | 7 - 8 | 805 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Detect Genomic DNA Mus musculus Probe spans exons |