Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400024477.mm.1 |
TC0400003043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024477.mm.1 |
NM_207685.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024477.mm.1 |
NM_207685.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024472.mm.1 |
TC0400003043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024472.mm.1 |
NM_001177883.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024472.mm.1 |
NM_001177883.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024445.mm.1 |
TC0400003043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024445.mm.1 |
NM_001177883.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024445.mm.1 |
NM_001177883.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024445.mm.1 |
NM_010486.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024445.mm.1 |
NM_010486.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024445.mm.1 |
NM_207685.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024445.mm.1 |
NM_207685.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024445.mm.1 |
NM_207686.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024445.mm.1 |
NM_207686.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024458.mm.1 |
TC0400003043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024458.mm.1 |
NM_010486.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024458.mm.1 |
NM_010486.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024458.mm.1 |
NM_207686.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024458.mm.1 |
NM_207686.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024469.mm.1 |
TC0400003043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024469.mm.1 |
NM_001177883.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024469.mm.1 |
NM_001177883.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024473.mm.1 |
TC0400003043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024473.mm.1 |
NM_001177883.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024473.mm.1 |
NM_001177883.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024476.mm.1 |
TC0400003043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024476.mm.1 |
NM_207685.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024476.mm.1 |
NM_207685.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024470.mm.1 |
TC0400003043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024470.mm.1 |
NM_001177883.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024470.mm.1 |
NM_001177883.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024471.mm.1 |
TC0400003043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024471.mm.1 |
NM_001177883.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024471.mm.1 |
NM_001177883.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024475.mm.1 |
TC0400003043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024475.mm.1 |
NM_207685.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024475.mm.1 |
NM_207685.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024446.mm.1 |
TC0400003043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024446.mm.1 |
NM_001177883.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024446.mm.1 |
NM_001177883.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024446.mm.1 |
NM_010486.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024446.mm.1 |
NM_010486.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024446.mm.1 |
NM_207685.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024446.mm.1 |
NM_207685.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024446.mm.1 |
NM_207686.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024446.mm.1 |
NM_207686.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024447.mm.1 |
TC0400003043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024447.mm.1 |
NM_001177883.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024447.mm.1 |
NM_001177883.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024447.mm.1 |
NM_010486.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024447.mm.1 |
NM_010486.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024447.mm.1 |
NM_207685.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024447.mm.1 |
NM_207685.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024447.mm.1 |
NM_207686.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024447.mm.1 |
NM_207686.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024448.mm.1 |
TC0400003043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024448.mm.1 |
NM_001177883.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024448.mm.1 |
NM_001177883.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024448.mm.1 |
NM_010486.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024448.mm.1 |
NM_010486.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024448.mm.1 |
NM_207685.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024448.mm.1 |
NM_207685.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024448.mm.1 |
NM_207686.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024448.mm.1 |
NM_207686.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024460.mm.1 |
TC0400003043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024460.mm.1 |
NM_010486.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024460.mm.1 |
NM_010486.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024460.mm.1 |
NM_207686.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024460.mm.1 |
NM_207686.1 |
Refseq |