Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myt1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001093775.1 | NP_001087244.1 | 20 - 21 | 3768 | 71 | |
NM_001093776.1 | NP_001087245.1 | 20 - 21 | 3674 | 71 | ||
NM_001093778.1 | NP_001087247.1 | 20 - 21 | 3759 | 71 | ||
NM_008666.3 | NP_032692.2 | 20 - 21 | 3762 | 71 | ||
GenBank mRNA | AB093283.1 | - | 17 - 18 | 3121 | 71 | |
AB212898.1 | - | 19 - 20 | 3366 | 71 | ||
AF004295.1 | - | 20 - 21 | 3578 | 71 | ||
BC079651.1 | - | 20 - 21 | 3643 | 71 | ||
BC094438.1 | - | 16 - 17 | 2999 | 30692780 | 71 | |
BC131677.1 | - | 19 - 20 | 3363 | 40111036 | 71 | |
U86338.1 | - | 20 - 21 | 3490 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |