Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myt1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001093775.1 | NP_001087244.1 | 7 - 8 | 997 | 85 | |
NM_001093776.1 | NP_001087245.1 | 7 - 8 | 909 | 85 | ||
NM_001093778.1 | NP_001087247.1 | 7 - 8 | 997 | 85 | ||
NM_008666.3 | NP_032692.2 | 7 - 8 | 997 | 85 | ||
GenBank mRNA | AB093283.1 | - | 4 - 5 | 356 | 85 | |
AB212898.1 | - | 6 - 7 | 595 | 85 | ||
AK049624.1 | - | 7 - 8 | 697 | 85 | ||
BC079651.1 | - | 7 - 8 | 878 | 85 | ||
BC094438.1 | - | 3 - 4 | 234 | 30692780 | 85 | |
BC131677.1 | - | 6 - 7 | 598 | 40111036 | 85 | |
U86338.1 | - | 7 - 8 | 716 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |