Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G2e3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015099.2 | NP_001015099.2 | 6 - 7 | 747 | 130 | |
NM_001167963.1 | NP_001161435.1 | 6 - 7 | 747 | 130 | ||
NM_001167964.1 | NP_001161436.1 | 6 - 7 | 747 | 130 | ||
GenBank mRNA | AK031333.1 | - | 6 - 7 | 607 | 130 | |
AK033513.1 | - | 6 - 7 | 689 | 130 | ||
AK084256.1 | - | 6 - 7 | 618 | 130 | ||
AK129332.1 | - | 6 - 7 | 650 | 130 | ||
BC086455.1 | - | 6 - 7 | 620 | 30458160 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |