Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meis3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC117532.1 | - | 5 - 6 | 525 | 40106322 | 78 |
BC118513.1 | - | 5 - 6 | 525 | 40106324 | 78 | |
U57344.1 | - | 5 - 6 | 580 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |