Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133234.2 | NP_573497.1 | 2 - 3 | 572 | 61 | |
GenBank mRNA | AF332560.1 | - | 2 - 3 | 517 | 61 | |
BC044782.2 | - | 2 - 3 | 539 | 61 | ||
BC060370.1 | - | 2 - 3 | 502 | 61 | ||
BC147261.1 | - | 2 - 3 | 400 | 9056294 | 61 | |
BC147262.1 | - | 2 - 3 | 400 | 9056313 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |