Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gltscr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081418.1 | NP_001074887.1 | 11 - 12 | 3538 | 91 | |
GenBank mRNA | AK220278.1 | - | 5 - 6 | 4211 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |