Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf7ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019426.2 | NP_062299.2 | 10 - 11 | 3092 | 62 | |
GenBank mRNA | AJ132702.1 | - | 10 - 11 | 3085 | 62 | |
AK132359.1 | - | 9 - 10 | 1712 | 62 | ||
AK137199.1 | - | 2 - 3 | 251 | 62 | ||
AK167542.1 | - | 11 - 12 | 3138 | 62 | ||
BC052460.1 | - | 10 - 11 | 3092 | 6834550 | 62 | |
BC060246.1 | - | 10 - 11 | 3075 | 5705945 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |